Learn More
MSC Fowler Carbide Ball Probe, 1 mm, 4 mm Stem, Tungsten Carbide
Catalog No. 5000529581
Encompass
Fowler Carbide Ball Probe is designed for precise measurements with height gages. It features a 1 mm tungsten carbide ball and a 4 mm stem, making it suitable for a variety of applications requiring high accuracy and durability.
  • Designed for precise measurements with height gages.
  • Features a 1 mm tungsten carbide ball for durability and accuracy.
  • Equipped with a 4 mm stem for broad compatibility with various height gage systems.
Catalog No. 50-005-29581 Supplier MSC Supplier No. 01005388
Fowler Carbide Ball Probe is designed for precise measurements with height gages. It features a 1 mm tungsten carbide ball and a 4 mm stem, making it suitable for a variety of applications requiring high accuracy and durability.
  • Designed for precise measurements with height gages.
  • Features a 1 mm tungsten carbide ball for durability and accuracy.
  • Equipped with a 4 mm stem for broad compatibility with various height gage systems.