Learn More
MSC Fowler Dial Bore Gage, 4-12 in, 0.0005 in Graduation, 5 Anvils
Catalog No. 5000531999
Encompass
This Dial Bore Gage measures diameters from 4 to 12 inches with a graduation of 0.0005 inches. It includes an indicator, extension holder, and comes with 5 carbide anvils and a wooden storage case, making it suitable for precise bore measurements in various applications.
  • Measures diameters from 4 to 12 inches
  • Features 0.0005 inch graduation
  • Includes indicator
  • Includes extension holder
  • Comes with wooden storage case
  • Equipped with 5 carbide anvils
  • Features chrome plated handle
  • Includes 2 extensions
Catalog No. 50-005-31999 Supplier MSC Supplier No. 70319850
This Dial Bore Gage measures diameters from 4 to 12 inches with a graduation of 0.0005 inches. It includes an indicator, extension holder, and comes with 5 carbide anvils and a wooden storage case, making it suitable for precise bore measurements in various applications.
  • Measures diameters from 4 to 12 inches
  • Features 0.0005 inch graduation
  • Includes indicator
  • Includes extension holder
  • Comes with wooden storage case
  • Equipped with 5 carbide anvils
  • Features chrome plated handle
  • Includes 2 extensions