Learn More
MSC Fowler Dial Bore Gage Set, 0.7-1.5 in Range, 0.0005 in Graduation, Carbide Anvils
Catalog No. 5000528098
Encompass
This dial bore gage set is designed for precise internal measurements, offering a range of 0.7 to 1.5 inches with a graduation of 0.0005 inches. It features durable carbide anvils for accurate readings.
  • Easily adjustable
  • Self-centering mechanism
  • Insulated grip for comfortable use
  • Chrome plated handle
  • Includes a fitted case for storage and transport
  • Comes with an indicator
  • Includes an extension holder
Catalog No. 50-005-28098 Supplier MSC Supplier No. 65002214
This dial bore gage set is designed for precise internal measurements, offering a range of 0.7 to 1.5 inches with a graduation of 0.0005 inches. It features durable carbide anvils for accurate readings.
  • Easily adjustable
  • Self-centering mechanism
  • Insulated grip for comfortable use
  • Chrome plated handle
  • Includes a fitted case for storage and transport
  • Comes with an indicator
  • Includes an extension holder