Learn More
MSC Fowler Dial Caliper, 12 inch, 0.001 inch Graduation, Stainless Steel
Catalog No. 5000529937
Encompass
This dial caliper is designed for precise measurements, offering a 12 inch maximum measurement with a 0.001 inch graduation. It features stainless steel jaws and is capable of performing outside, inside, step, and depth measurements.
  • Measures outside, step, inside, and depth.
  • Longer jaw design for extended reach.
  • Constructed with stainless steel jaws for durability.
  • Provides an accuracy of +/-0.0020 "
  • Features a range of 0.1000 inch per revolution.
Catalog No. 50-005-29937 Supplier MSC Supplier No. 06418222
This dial caliper is designed for precise measurements, offering a 12 inch maximum measurement with a 0.001 inch graduation. It features stainless steel jaws and is capable of performing outside, inside, step, and depth measurements.
  • Measures outside, step, inside, and depth.
  • Longer jaw design for extended reach.
  • Constructed with stainless steel jaws for durability.
  • Provides an accuracy of +/-0.0020 "
  • Features a range of 0.1000 inch per revolution.