Learn More
MSC Fowler Dial Caliper, 6 inch, 0.001 inch Graduation, Shockproof
Catalog No. 5000527519
Encompass
This dial caliper is designed for precise measurement of outside, step, inside, and depth dimensions. It features a shockproof design for enhanced durability and stainless steel construction for reliable performance. The caliper has a white dial face and a gray dial case.
  • Measures outside, inside, step, and depth dimensions
  • Maximum measurement of 6 inch
  • Graduation of 0.001 inch
  • Accuracy of +/-0.001 inch
  • Shockproof design
  • Stainless steel construction
  • Stainless steel jaws
  • White dial face
  • Gray dial case
Catalog No. 50-005-27519 Supplier MSC Supplier No. 85213437
This dial caliper is designed for precise measurement of outside, step, inside, and depth dimensions. It features a shockproof design for enhanced durability and stainless steel construction for reliable performance. The caliper has a white dial face and a gray dial case.
  • Measures outside, inside, step, and depth dimensions
  • Maximum measurement of 6 inch
  • Graduation of 0.001 inch
  • Accuracy of +/-0.001 inch
  • Shockproof design
  • Stainless steel construction
  • Stainless steel jaws
  • White dial face
  • Gray dial case