Learn More
MSC Fowler Dial Height Gage, 12 in, 0.001 in Fine Indicator Reading, 0.0015 in Accuracy
Catalog No. 5000536731
Encompass
Fowler dial height gages are designed for precise measurement in various applications. This 12 in model features both fine and coarse readings, along with a dial and counter display for clear, accurate results. Its resettable digital counter allows for individual upward and downward measurements from a preset zero point.
  • 0.001 in fine indicator reading
  • 0.1 in coarse reading graduation
  • Dial and counter display for clear readings
  • Resettable digital counter with two displays that individually measure upward and downward measurements from a preset zero.
  • Accurate to 0.0015 in
Catalog No. 50-005-36731 Supplier MSC Supplier No. 93296580
Fowler dial height gages are designed for precise measurement in various applications. This 12 in model features both fine and coarse readings, along with a dial and counter display for clear, accurate results. Its resettable digital counter allows for individual upward and downward measurements from a preset zero point.
  • 0.001 in fine indicator reading
  • 0.1 in coarse reading graduation
  • Dial and counter display for clear readings
  • Resettable digital counter with two displays that individually measure upward and downward measurements from a preset zero.
  • Accurate to 0.0015 in