Learn More
MSC Fowler Dial Indicator & Base Kit, 0.001 in Graduation, 0-100 Dial, Black Dial
Catalog No. 5000513049
Encompass
This dial indicator and base kit features a 0.001 in graduation and a 0-100 black dial. It includes a magnetic base and a premium indicator, making it suitable for various precision measurement applications.
  • Dial indicator type
  • Magnetic base style
  • 0.001 in graduation
  • Black dial color
  • 0-100 dial reading
  • 1.0000 in measuring range
  • Includes mag base and premium indicator
  • Holder type A
Catalog No. 50-005-13049 Supplier MSC Supplier No. 77751451
This dial indicator and base kit features a 0.001 in graduation and a 0-100 black dial. It includes a magnetic base and a premium indicator, making it suitable for various precision measurement applications.
  • Dial indicator type
  • Magnetic base style
  • 0.001 in graduation
  • Black dial color
  • 0-100 dial reading
  • 1.0000 in measuring range
  • Includes mag base and premium indicator
  • Holder type A