Learn More
MSC Fowler Dial Indicator & Base Kit, 1.0000 in, 0.0010 in Accuracy
Catalog No. 5000514480
Encompass
This Dial Indicator and Base Kit is designed for precise measurements. It features a 2.3000 in base length, 1.8000 in base height, 2 in base width, a 1.0000 in maximum measurement range, a 0-100 dial, and 0.0010 in accuracy.
  • Sealed back plate.
  • Fitted case included.
  • Indicator included.
  • Magnetic base included.
  • Water resistant indicator.
Catalog No. 50-005-14480 Supplier MSC Supplier No. 56066830
This Dial Indicator and Base Kit is designed for precise measurements. It features a 2.3000 in base length, 1.8000 in base height, 2 in base width, a 1.0000 in maximum measurement range, a 0-100 dial, and 0.0010 in accuracy.
  • Sealed back plate.
  • Fitted case included.
  • Indicator included.
  • Magnetic base included.
  • Water resistant indicator.