Learn More
MSC Fowler Dial Indicator Kit, 0.0600 in Max, 0.000500 in Grad
Catalog No. 5000514398
Encompass
This kit includes a dial indicator and a base for precise measurement applications. It features a horizontal test indicator and a fine adjustment attachment, making it suitable for detailed inspections and readings.
  • Maximum measurement of 0.0600 in
  • Graduation of 0.000500 in
  • White dial color
  • Dial reading of 0-15-0
  • Includes horizontal test indicator
  • Includes fine adjustment attachment
  • Features a 1-1/2 in dial diameter
Catalog No. 50-005-14398 Supplier MSC Supplier No. 77751410
This kit includes a dial indicator and a base for precise measurement applications. It features a horizontal test indicator and a fine adjustment attachment, making it suitable for detailed inspections and readings.
  • Maximum measurement of 0.0600 in
  • Graduation of 0.000500 in
  • White dial color
  • Dial reading of 0-15-0
  • Includes horizontal test indicator
  • Includes fine adjustment attachment
  • Features a 1-1/2 in dial diameter