Learn More
MSC Fowler Dial Test Indicator Kit, 0.060 in Max, 0.0004 in Accuracy, 12 Piece
Catalog No. 5000535812
Encompass
This dial test indicator kit is designed for precise measurement applications. This 12-piece kit features a horizontal indicator style with a maximum measurement of 0.060 inches and an accuracy of 0.0004 inches. It is suitable for various shop and laboratory settings.
  • Jeweled movement
  • Easy-to-read tilted dial
  • Automatic reversal
  • Revolution counter
  • 210° swivel arc
  • Axial support for stable measurements
  • Shop-hardened case for durability
  • Includes contact point case, two dovetail stems, contact point wrench, height gage bar with clamp, three contact points, and an adapter sleeve
Catalog No. 50-005-35812 Supplier MSC Supplier No. 81901571
This dial test indicator kit is designed for precise measurement applications. This 12-piece kit features a horizontal indicator style with a maximum measurement of 0.060 inches and an accuracy of 0.0004 inches. It is suitable for various shop and laboratory settings.
  • Jeweled movement
  • Easy-to-read tilted dial
  • Automatic reversal
  • Revolution counter
  • 210° swivel arc
  • Axial support for stable measurements
  • Shop-hardened case for durability
  • Includes contact point case, two dovetail stems, contact point wrench, height gage bar with clamp, three contact points, and an adapter sleeve