Learn More
MSC Fowler Digital Electronic Drop Indicator, 1/2 in, 0.0005 in Resolution
Catalog No. 5000535806
Encompass
This electronic drop indicator features a digital display for clear readings and a lug back for secure attachment. Designed for versatility, it measures in both inch and metric units and comes equipped with a durable carbide tipped point. The indicator's face can rotate up to 270 degrees, allowing for flexible viewing angles during operation.
  • Digital display for clear readings
  • Lug back type for secure mounting
  • Measures in both inch and metric units
  • Solid carbide contact point for enhanced durability
  • Face rotates up to 270 degrees for flexible viewing
  • Powered by a CR2032 battery
  • Resolution of 0.0005 in (0.01 mm)
  • Measurement range of 0 to 1/2 in (0 to 12.5 mm)
  • Stem diameter of 3/8 in
Catalog No. 50-005-35806 Supplier MSC Supplier No. 79401493
This electronic drop indicator features a digital display for clear readings and a lug back for secure attachment. Designed for versatility, it measures in both inch and metric units and comes equipped with a durable carbide tipped point. The indicator's face can rotate up to 270 degrees, allowing for flexible viewing angles during operation.
  • Digital display for clear readings
  • Lug back type for secure mounting
  • Measures in both inch and metric units
  • Solid carbide contact point for enhanced durability
  • Face rotates up to 270 degrees for flexible viewing
  • Powered by a CR2032 battery
  • Resolution of 0.0005 in (0.01 mm)
  • Measurement range of 0 to 1/2 in (0 to 12.5 mm)
  • Stem diameter of 3/8 in