Learn More
MSC Fowler Electronic Blade Micrometer, 25-50 mm, 0.001 mm Resolution, IP54
Catalog No. 5000513398
Encompass
This electronic blade micrometer can be used for measuring the outside diameters of work pieces with narrow grooves, T-slots, and other dimensions blocked by shoulders or lands inaccessible by standard micrometers.
  • Water resistant to IP54 specifications
  • True inch/metric conversion
  • Direct RS232 output
  • Large easy-to-read LCD display
  • Includes shop-hardened case
  • Includes standard for sizes over 1 inch
Catalog No. 50-005-13398 Supplier MSC Supplier No. 05022892
This electronic blade micrometer can be used for measuring the outside diameters of work pieces with narrow grooves, T-slots, and other dimensions blocked by shoulders or lands inaccessible by standard micrometers.
  • Water resistant to IP54 specifications
  • True inch/metric conversion
  • Direct RS232 output
  • Large easy-to-read LCD display
  • Includes shop-hardened case
  • Includes standard for sizes over 1 inch