Learn More
MSC Fowler Electronic Caliper, 0-1.25 in / 0-31.5 mm, 0.0005 in / 0.01 mm Resolution, Hardened Stainless Steel
Catalog No. 5000512289
Encompass
This electronic caliper is a precision tool designed for accurate measurements. Constructed from hardened stainless steel, it is built for heavy-duty use and provides reliable readings. It features spring-loaded travel for consistent positioning and offers direct inch, metric, and fraction conversion.
  • Inch, metric, and fraction reading
  • Ideal for measuring a wide range of materials
  • Spring-loaded travel
  • Wide-faced flat jaw for easy positioning
  • Direct inch/metric/fraction conversion
  • Measuring speed of 60 in/second
  • Large, high-contrast LCD display
  • Hanging loop included
  • Includes case
  • Batteries included
  • Hardened stainless steel construction
  • Application for outside diameter
  • CR2032 lithium battery power source
Catalog No. 50-005-12289 Supplier MSC Supplier No. 66024555
This electronic caliper is a precision tool designed for accurate measurements. Constructed from hardened stainless steel, it is built for heavy-duty use and provides reliable readings. It features spring-loaded travel for consistent positioning and offers direct inch, metric, and fraction conversion.
  • Inch, metric, and fraction reading
  • Ideal for measuring a wide range of materials
  • Spring-loaded travel
  • Wide-faced flat jaw for easy positioning
  • Direct inch/metric/fraction conversion
  • Measuring speed of 60 in/second
  • Large, high-contrast LCD display
  • Hanging loop included
  • Includes case
  • Batteries included
  • Hardened stainless steel construction
  • Application for outside diameter
  • CR2032 lithium battery power source