Learn More
MSC Fowler Electronic Caliper Gage, 6.000 in, 0.0100 in Resolution, Spring-loaded
Catalog No. 5000536854
Encompass
This electronic caliper gage from Fowler offers precise measurements with a resolution of 0.0100 inch and a maximum measurement capability of 6.000 inches. It is spring-loaded for ease of use and operates on a CR2032 battery. The gage comes complete with a protective case.
  • Spring-loaded design for easy operation.
  • Comes with a protective case for storage.
  • Operates on a CR2032 battery.
  • Measures up to 6.000 inches.
  • Provides a resolution of 0.0100.
  • No data output feature.
Catalog No. 50-005-36854 Supplier MSC Supplier No. 42314708
This electronic caliper gage from Fowler offers precise measurements with a resolution of 0.0100 inch and a maximum measurement capability of 6.000 inches. It is spring-loaded for ease of use and operates on a CR2032 battery. The gage comes complete with a protective case.
  • Spring-loaded design for easy operation.
  • Comes with a protective case for storage.
  • Operates on a CR2032 battery.
  • Measures up to 6.000 inches.
  • Provides a resolution of 0.0100.
  • No data output feature.