Learn More
MSC Fowler Electronic Caliper Gage, External, 0.790-1.6 in Range, 0.0002 in Resolution, ±0.0008 in Accuracy
Catalog No. 5000533401
Encompass
This electronic caliper gage provides quick and accurate external measurements. It features a range of 0.790-1.6 in (20-40 mm), a resolution of 0.0002 in (0.005 mm), and an accuracy of ±0.0008 in (±0.02 mm), suitable for various industrial applications.
  • Easy to hold
  • Rugged housing
  • Large, LCD display
  • Preset capability
  • Direct inch/metric conversion
  • Absolute/incremental measurements
  • Includes case
Catalog No. 50-005-33401 Supplier MSC Supplier No. 05022611
This electronic caliper gage provides quick and accurate external measurements. It features a range of 0.790-1.6 in (20-40 mm), a resolution of 0.0002 in (0.005 mm), and an accuracy of ±0.0008 in (±0.02 mm), suitable for various industrial applications.
  • Easy to hold
  • Rugged housing
  • Large, LCD display
  • Preset capability
  • Direct inch/metric conversion
  • Absolute/incremental measurements
  • Includes case