Learn More
MSC Fowler Electronic Caliper Gage, Inside, 12.70 to 150 mm
Catalog No. 5000531198
Encompass
This electronic caliper gage is designed for inside measurements, offering a range of 12.70 mm to 150 mm with 0.01 inches accuracy. It features a Radius to Radius contact point, is powered by a CR2032 battery, and supports absolute/incremental modes with RS-232 output.
  • Measures inside dimensions for hole, groove, and tube thicknesses.
  • Digital LCD displays inch and metric units.
  • Absolute and incremental modes with preset reference storage.
  • Spring-loaded design for consistent measuring force.
  • Thumb lever for movable point retraction.
  • RS-232 output for data transfer.
  • Durable construction: plastic housing, steel arms, hardened steel contact points.
  • Includes 3V CR2032 battery.
  • Comes with aluminum case and foam inserts.
Catalog No. 50-005-31198 Supplier MSC Supplier No. 79437356
This electronic caliper gage is designed for inside measurements, offering a range of 12.70 mm to 150 mm with 0.01 inches accuracy. It features a Radius to Radius contact point, is powered by a CR2032 battery, and supports absolute/incremental modes with RS-232 output.
  • Measures inside dimensions for hole, groove, and tube thicknesses.
  • Digital LCD displays inch and metric units.
  • Absolute and incremental modes with preset reference storage.
  • Spring-loaded design for consistent measuring force.
  • Thumb lever for movable point retraction.
  • RS-232 output for data transfer.
  • Durable construction: plastic housing, steel arms, hardened steel contact points.
  • Includes 3V CR2032 battery.
  • Comes with aluminum case and foam inserts.