Learn More
MSC Fowler Electronic Caliper, IP67, 0-6 in Range, Stainless steel
Catalog No. 5000515804
Encompass
This electronic caliper offers a 0-6 in (150 mm) measurement range and is protected to IP67 standards. Constructed from stainless steel, it provides accurate measurements for inside, outside, depth, and step applications, featuring data output capabilities.
  • Equipped with a new HID interface, eliminating the need for a dongle.
  • Includes an extra battery tray and 1 in standard.
  • Supplied with NIST certification for traceability.
  • Features a heavy duty frame for 4-way measuring.
  • Designed with a super large display for easy reading.
  • Offers Bluetooth connectivity for data output.
  • Includes inside diameter jaws and a depth measurement rod.
  • Comes with a protective case and battery.
Catalog No. 50-005-15804 Supplier MSC Supplier No. 97898118
This electronic caliper offers a 0-6 in (150 mm) measurement range and is protected to IP67 standards. Constructed from stainless steel, it provides accurate measurements for inside, outside, depth, and step applications, featuring data output capabilities.
  • Equipped with a new HID interface, eliminating the need for a dongle.
  • Includes an extra battery tray and 1 in standard.
  • Supplied with NIST certification for traceability.
  • Features a heavy duty frame for 4-way measuring.
  • Designed with a super large display for easy reading.
  • Offers Bluetooth connectivity for data output.
  • Includes inside diameter jaws and a depth measurement rod.
  • Comes with a protective case and battery.