Learn More
MSC Fowler Electronic Caliper, IP67, 300 mm, 0.01 mm Resolution, Bluetooth
Catalog No. 5000510209
Encompass
The Fowler Electronic Caliper is designed for precise measurements in demanding environments. Featuring an IP67 rating for protection against dust and water, it offers a measurement range of 0-300 mm with a resolution of 0.01 mm. This caliper includes Bluetooth connectivity for easy data output, making it suitable for various laboratory and industrial applications.
  • Measures depth, inside diameter, outside diameter, and step
  • Includes inside diameter jaws
  • Includes depth measurement rod
  • Comes with a case and battery
  • Features a new HID interface for data output, no dongle needed
  • Includes an extra battery tray and 1 inch standard
  • Supplied with NIST certification
  • Heavy-duty frame with four-way measuring capabilities
  • Equipped with a super large display
  • Protected to IP67 standards
Catalog No. 50-005-10209 Supplier MSC Supplier No. 96050703
The Fowler Electronic Caliper is designed for precise measurements in demanding environments. Featuring an IP67 rating for protection against dust and water, it offers a measurement range of 0-300 mm with a resolution of 0.01 mm. This caliper includes Bluetooth connectivity for easy data output, making it suitable for various laboratory and industrial applications.
  • Measures depth, inside diameter, outside diameter, and step
  • Includes inside diameter jaws
  • Includes depth measurement rod
  • Comes with a case and battery
  • Features a new HID interface for data output, no dongle needed
  • Includes an extra battery tray and 1 inch standard
  • Supplied with NIST certification
  • Heavy-duty frame with four-way measuring capabilities
  • Equipped with a super large display
  • Protected to IP67 standards