Learn More
MSC Fowler Electronic Caliper, Polycarbonate, 6 inch Range, 0.0005 inch Resolution
Catalog No. 5000530306
Encompass
This electronic caliper offers precise measurements with a 6 inch / 150 mm range and a resolution of 0.0005 inch / 0.01 mm. Constructed with a polycarbonate frame, it provides 4-way measuring capabilities for depth, inside and outside diameters, and step measurements. It features direct inch/metric/fraction conversion and is designed for accuracy in various applications.
  • Polycarbonate frame for lightweight and durable use.
  • Large, easy-to-read display.
  • Absolute and incremental measurement modes.
  • Direct inch, metric, and fraction conversion.
  • Measures depth, inside diameter, outside diameter, and step.
  • IP67 rated for dust and water resistance.
  • Powered by a CR2032 lithium battery, included with the device.
  • Comes with a clip-on vinyl pouch for storage.
Catalog No. 50-005-30306 Supplier MSC Supplier No. 74040239
This electronic caliper offers precise measurements with a 6 inch / 150 mm range and a resolution of 0.0005 inch / 0.01 mm. Constructed with a polycarbonate frame, it provides 4-way measuring capabilities for depth, inside and outside diameters, and step measurements. It features direct inch/metric/fraction conversion and is designed for accuracy in various applications.
  • Polycarbonate frame for lightweight and durable use.
  • Large, easy-to-read display.
  • Absolute and incremental measurement modes.
  • Direct inch, metric, and fraction conversion.
  • Measures depth, inside diameter, outside diameter, and step.
  • IP67 rated for dust and water resistance.
  • Powered by a CR2032 lithium battery, included with the device.
  • Comes with a clip-on vinyl pouch for storage.