Learn More
MSC Fowler Gage Block Set, 0.05-4 inch, 36 Piece, Economy, Steel
Catalog No. 5000512512
Encompass
This gage block set is a comprehensive 36-piece collection designed for precise measurements. Constructed from steel with an economy accuracy grade, this set provides a measurement range from 0.05 to 4 inches. It comes with NIST traceability certification, ensuring reliability for various applications.
  • Includes 36 pieces for versatile measurement tasks.
  • Constructed from durable steel for long-lasting performance.
  • Features an economy accuracy grade suitable for general precision work.
  • Offers a broad measurement range of 0.05 to 4 inches.
  • Comes with NIST traceability certification for assured accuracy.
  • Provided in a fitted case for organized storage and protection.
Catalog No. 50-005-12512 Supplier MSC Supplier No. 67260224
This gage block set is a comprehensive 36-piece collection designed for precise measurements. Constructed from steel with an economy accuracy grade, this set provides a measurement range from 0.05 to 4 inches. It comes with NIST traceability certification, ensuring reliability for various applications.
  • Includes 36 pieces for versatile measurement tasks.
  • Constructed from durable steel for long-lasting performance.
  • Features an economy accuracy grade suitable for general precision work.
  • Offers a broad measurement range of 0.05 to 4 inches.
  • Comes with NIST traceability certification for assured accuracy.
  • Provided in a fitted case for organized storage and protection.