Learn More
MSC Fowler Indicator Set, X-Proof, Water Resistant, 1 inch, 0.001 inch
Catalog No. 5000531560
Encompass
This X-Proof Water Resistant Indicator Set is a robust, shockproof measurement tool for precision in demanding environments. It features an IP54-rated X-Proof indicator with 1 in travel and 0.001 in reading, plus a magnetic base for stable positioning. It ensures accurate, reliable measurements with enhanced water and dust protection.
  • Robust, shockproof construction.
  • IP54 X-Proof indicator.
  • 1 in travel and 0.001 in reading.
  • Magnetic base with fine adjustment.
  • Base features 85 lb pull magnet with on-off lever.
  • V-base for stable placement on round surfaces.
  • Rubber bellows on stem and O-ring seals for water resistance.
  • Includes sealed back plate.
Catalog No. 50-005-31560 Supplier MSC Supplier No. 65861379
This X-Proof Water Resistant Indicator Set is a robust, shockproof measurement tool for precision in demanding environments. It features an IP54-rated X-Proof indicator with 1 in travel and 0.001 in reading, plus a magnetic base for stable positioning. It ensures accurate, reliable measurements with enhanced water and dust protection.
  • Robust, shockproof construction.
  • IP54 X-Proof indicator.
  • 1 in travel and 0.001 in reading.
  • Magnetic base with fine adjustment.
  • Base features 85 lb pull magnet with on-off lever.
  • V-base for stable placement on round surfaces.
  • Rubber bellows on stem and O-ring seals for water resistance.
  • Includes sealed back plate.