Learn More
MSC Fowler Indicator Transfer Stand, Fine Adjustment, Steel, 7 in Post Height
Catalog No. 5000527579
Encompass
This indicator transfer stand features a fine adjustment arm, making it suitable for precise measurements and transfers. Constructed from durable steel, it provides a stable base for various laboratory and workshop applications.
  • Fine adjustment arm for precise positioning
  • Durable steel construction
  • Includes a holder
  • Rectangular base for enhanced stability
  • Base dimensions of 2.2000 in length and 1.5000 in width
  • Post height of 7 in
Catalog No. 50-005-27579 Supplier MSC Supplier No. 76646413
This indicator transfer stand features a fine adjustment arm, making it suitable for precise measurements and transfers. Constructed from durable steel, it provides a stable base for various laboratory and workshop applications.
  • Fine adjustment arm for precise positioning
  • Durable steel construction
  • Includes a holder
  • Rectangular base for enhanced stability
  • Base dimensions of 2.2000 in length and 1.5000 in width
  • Post height of 7 in