Learn More
MSC Fowler Inside Micrometer Set, 2-6 in, 0.001 in Graduations, Carbide Faces
Catalog No. 5000529702
Encompass
This mechanical inside micrometer set offers a 2 to 6 inch measurement range, featuring an inside micrometer head and hardened, lapped extension rods. It is equipped with durable carbide measuring faces, and its hardened, precision-ground anvil and spindle with microlap finish ensure accurate readings and easy zero adjustment.
  • 2 to 6 inch measurement range
  • 0.001 inch graduations
  • Carbide measuring faces
  • Hardened and precision ground anvil and spindle with microlap finish
  • Quick and easy zero adjustment on sleeve
  • Friction thimble
  • Hardened spindle thread
  • Positive locking nut on spindle
  • Satin chrome finish
  • Includes fitted case, wrench, and standard
Catalog No. 50-005-29702 Supplier MSC Supplier No. 06399307
This mechanical inside micrometer set offers a 2 to 6 inch measurement range, featuring an inside micrometer head and hardened, lapped extension rods. It is equipped with durable carbide measuring faces, and its hardened, precision-ground anvil and spindle with microlap finish ensure accurate readings and easy zero adjustment.
  • 2 to 6 inch measurement range
  • 0.001 inch graduations
  • Carbide measuring faces
  • Hardened and precision ground anvil and spindle with microlap finish
  • Quick and easy zero adjustment on sleeve
  • Friction thimble
  • Hardened spindle thread
  • Positive locking nut on spindle
  • Satin chrome finish
  • Includes fitted case, wrench, and standard