Learn More
MSC Fowler Machinist Caliper & Micrometer Kit, 4 pc, Electronic
Catalog No. 5000511856
Encompass
This Machinist Caliper & Micrometer Kit is a comprehensive 4-piece set designed for precision measurement tasks in various applications.
  • Includes an electronic caliper, micrometer, indicator, and magnetic base.
  • The electronic caliper features a resolution of 0.000500 in.
Catalog No. 50-005-11856 Supplier MSC Supplier No. 77750719
This Machinist Caliper & Micrometer Kit is a comprehensive 4-piece set designed for precision measurement tasks in various applications.
  • Includes an electronic caliper, micrometer, indicator, and magnetic base.
  • The electronic caliper features a resolution of 0.000500 in.