Learn More
MSC Fowler Mechanical Micrometer Set, 3 Pc, 0-3 in, Carbide Face
Catalog No. 5000512432
Encompass
The Fowler 52-215-003-1 Mechanical Outside Micrometer Set is a comprehensive 3-piece set designed for precise external measurements from 0 to 3 inches. Each micrometer features a durable satin chrome finish and measuring faces crafted from solid carbide, ensuring longevity and accuracy. The set is equipped with both friction and ratchet stop thimbles, providing consistent measuring force for reliable results.
  • 3-piece set for a versatile range of measurements
  • Measurement range of 0 to 3 inches for various applications
  • Equipped with friction and ratchet stop thimbles for consistent force
  • Measuring faces are made from solid carbide for durability and accuracy
  • Features a satin chrome finish for enhanced resistance to wear
  • Includes a protective case and a wrench for convenience
Catalog No. 50-005-12432 Supplier MSC Supplier No. 06400832
The Fowler 52-215-003-1 Mechanical Outside Micrometer Set is a comprehensive 3-piece set designed for precise external measurements from 0 to 3 inches. Each micrometer features a durable satin chrome finish and measuring faces crafted from solid carbide, ensuring longevity and accuracy. The set is equipped with both friction and ratchet stop thimbles, providing consistent measuring force for reliable results.
  • 3-piece set for a versatile range of measurements
  • Measurement range of 0 to 3 inches for various applications
  • Equipped with friction and ratchet stop thimbles for consistent force
  • Measuring faces are made from solid carbide for durability and accuracy
  • Features a satin chrome finish for enhanced resistance to wear
  • Includes a protective case and a wrench for convenience