Learn More
MSC Fowler Mechanical Outside Micrometer, 1 in Range, 0.0001 in Graduation, Friction Thimble, Solid Carbide Faces
Catalog No. 5000529715
Encompass
This mechanical outside micrometer provides precise external measurements with a 1 in range and 0.0001 in graduation. It features a friction thimble and solid carbide measuring faces, ensuring accuracy and durability. The design includes quick and easy zero adjustment on the sleeve for convenient use.
  • Precise external measurements
  • 1 inch maximum measurement range
  • 0.0001 inch graduation for high accuracy
  • Friction thimble type
  • Solid carbide measuring faces
  • Quick and easy zero adjustment on sleeve
  • Includes protective case and wrench
Catalog No. 50-005-29715 Supplier MSC Supplier No. 06400618
This mechanical outside micrometer provides precise external measurements with a 1 in range and 0.0001 in graduation. It features a friction thimble and solid carbide measuring faces, ensuring accuracy and durability. The design includes quick and easy zero adjustment on the sleeve for convenient use.
  • Precise external measurements
  • 1 inch maximum measurement range
  • 0.0001 inch graduation for high accuracy
  • Friction thimble type
  • Solid carbide measuring faces
  • Quick and easy zero adjustment on sleeve
  • Includes protective case and wrench