Learn More
MSC Fowler Mechanical Outside Micrometer, 3 in Range, 0.0001 in Graduation, Ratchet Stop Thimble
Catalog No. 5000516895
Encompass
The mechanical outside micrometer offers a 3 in range and 0.0001 in graduation for precise measurements. It is equipped with a ratchet stop thimble and provides an accuracy of 0.000230 in.
  • Quick, easy zero adjustment on sleeve
  • Measuring face material is solid carbide
  • Includes wrench and case
  • Does not feature a rotating spindle
  • Does not include a digital counter
Catalog No. 50-005-16895 Supplier MSC Supplier No. 06400634
The mechanical outside micrometer offers a 3 in range and 0.0001 in graduation for precise measurements. It is equipped with a ratchet stop thimble and provides an accuracy of 0.000230 in.
  • Quick, easy zero adjustment on sleeve
  • Measuring face material is solid carbide
  • Includes wrench and case
  • Does not feature a rotating spindle
  • Does not include a digital counter