Learn More
MSC Fowler Mechanical Outside Micrometer, 6 in, 0.0001 in, Ratchet Stop
Catalog No. 5000514038
Encompass
This mechanical outside micrometer is designed for precise measurements up to 6 inches, with a graduation of 0.0001 inch and an accuracy of 0.000250 " It features a ratchet stop thimble for consistent measuring force and solid carbide measuring faces for durability.
  • Quick, easy zero adjustment on sleeve
  • Includes wrench
  • Includes case
Catalog No. 50-005-14038 Supplier MSC Supplier No. 06400667
This mechanical outside micrometer is designed for precise measurements up to 6 inches, with a graduation of 0.0001 inch and an accuracy of 0.000250 " It features a ratchet stop thimble for consistent measuring force and solid carbide measuring faces for durability.
  • Quick, easy zero adjustment on sleeve
  • Includes wrench
  • Includes case