Learn More
MSC Fowler Mechanical Outside Micrometer Set, 6 Pc, 6 to 12 in Measurement, Ratchet Stop Thimble, Solid Carbide Measuring Face
Catalog No. 5000529204
Encompass
This mechanical outside micrometer set is designed for precise external measurements. It includes 6 pieces and covers a measurement range from 6 to 12 inches. The set features a ratchet stop thimble for consistent measuring force and solid carbide measuring faces for durability and accuracy.
  • Ratchet stop thimble ensures consistent measuring pressure.
  • Solid carbide measuring faces provide durability and accurate readings.
  • Contains 6 pieces for various measurement needs.
  • Measurement range from 6 to 12 inches for broad application.
  • Features a satin chrome finish for enhanced durability.
  • Includes a wrench for adjustments and a protective case for storage.
Catalog No. 50-005-29204 Supplier MSC Supplier No. 06400873
This mechanical outside micrometer set is designed for precise external measurements. It includes 6 pieces and covers a measurement range from 6 to 12 inches. The set features a ratchet stop thimble for consistent measuring force and solid carbide measuring faces for durability and accuracy.
  • Ratchet stop thimble ensures consistent measuring pressure.
  • Solid carbide measuring faces provide durability and accurate readings.
  • Contains 6 pieces for various measurement needs.
  • Measurement range from 6 to 12 inches for broad application.
  • Features a satin chrome finish for enhanced durability.
  • Includes a wrench for adjustments and a protective case for storage.