Learn More
MSC Fowler Outside Micrometer Set, 0-4 inch, Ratchet Stop, Solid Carbide
Catalog No. 5000532001
Encompass
This outside micrometer set provides precise measurements for various applications. It features a clear digital counter for easy readings across a 0-4 inch range, a ratchet stop thimble, and solid carbide measuring faces. Ideal for precision machining, quality control, and inspection.
  • Digital counter for clear readings
  • Resolution of 0.0001 inch
  • Ratchet stop thimble
  • Solid carbide measuring faces
  • Microfine graduations on satin chrome
  • Positive locking nut on spindle
  • Includes standards and case
Catalog No. 50-005-32001 Supplier MSC Supplier No. 72286354
This outside micrometer set provides precise measurements for various applications. It features a clear digital counter for easy readings across a 0-4 inch range, a ratchet stop thimble, and solid carbide measuring faces. Ideal for precision machining, quality control, and inspection.
  • Digital counter for clear readings
  • Resolution of 0.0001 inch
  • Ratchet stop thimble
  • Solid carbide measuring faces
  • Microfine graduations on satin chrome
  • Positive locking nut on spindle
  • Includes standards and case