Learn More
Andwin Scientific FOWLER PERCISION
SDP
Catalog No. NC3761524
Encompass
  • Bluetooth connectivity for data transfer
  • Pistol grip handle for consistent measuring pressure
  • Self-centering heads for easy alignment
  • Measured range of 0.080 in - 0.250 in / 2 mm - 6 mm
  • Resolution of 0.00005 in / 0.001 mm
  • Accuracy of ±0.00015 in / 0.0038 mm
  • Large, easy-to-read swivel display
  • Includes 5 heads, 1 handle, and 3 setting rings
  • Adjustable trigger travel for reduced hand fatigue
  • Fixed, non-removable anvils for stability
  • Blind bore capability for larger heads
  • UKAS certified components for quality assurance
  • 5-year warranty for peace of mind
Catalog No. NC3761524 Supplier Andwin Scientific Supplier No. 54567009BT
Add to Cart
Add to Cart
Request a Quote
  • Bluetooth connectivity for data transfer
  • Pistol grip handle for consistent measuring pressure
  • Self-centering heads for easy alignment
  • Measured range of 0.080 in - 0.250 in / 2 mm - 6 mm
  • Resolution of 0.00005 in / 0.001 mm
  • Accuracy of ±0.00015 in / 0.0038 mm
  • Large, easy-to-read swivel display
  • Includes 5 heads, 1 handle, and 3 setting rings
  • Adjustable trigger travel for reduced hand fatigue
  • Fixed, non-removable anvils for stability
  • Blind bore capability for larger heads
  • UKAS certified components for quality assurance
  • 5-year warranty for peace of mind