Learn More
MSC Fowler Proximity Cable, USB Connection, RS-232 Output, CE Compliant
Catalog No. 5000528172
Encompass
This proximity cable provides RS-232 output via a USB connection, designed to expand the capabilities of your measurement devices. It is compatible with Sylvac calipers and Fowler Ultra-Cal V devices, ensuring reliable data transfer. The cable is also Conformite Europeenne (CE) compliant.
  • Provides RS-232 output with USB connection
  • Conformite Europeenne (CE) compliant
  • Enhances data transfer for compatible calipers
  • Expands functionality for Ultra-Cal V devices
Catalog No. 50-005-28172 Supplier MSC Supplier No. 52562808
This proximity cable provides RS-232 output via a USB connection, designed to expand the capabilities of your measurement devices. It is compatible with Sylvac calipers and Fowler Ultra-Cal V devices, ensuring reliable data transfer. The cable is also Conformite Europeenne (CE) compliant.
  • Provides RS-232 output with USB connection
  • Conformite Europeenne (CE) compliant
  • Enhances data transfer for compatible calipers
  • Expands functionality for Ultra-Cal V devices