Learn More
MSC Fowler Test Indicator Kit, 12 Piece, 0.060 in Max, 0.0004 in Accuracy
Catalog No. 5000531204
Encompass
The Fowler Test Indicator Kit offers precise measurement capabilities for diverse applications. This 12-piece kit features a horizontal indicator with a maximum measurement range of 0.060 inch and an accuracy of 0.0004 " It is designed for use in demanding laboratory and industrial environments.
  • Jeweled movement
  • Easy-to-read tilted dial
  • Axial support
  • 210° swivel arc
  • Automatic reversal
  • Revolution counter
  • Shop hardened case
  • Includes: dial test indicator, shop-hardened case, axial support, contact point case, two dovetail stems, contact point wrench, height gage bar with clamp, three contact points, adapter sleeve.
Catalog No. 50-005-31204 Supplier MSC Supplier No. 81901563
The Fowler Test Indicator Kit offers precise measurement capabilities for diverse applications. This 12-piece kit features a horizontal indicator with a maximum measurement range of 0.060 inch and an accuracy of 0.0004 " It is designed for use in demanding laboratory and industrial environments.
  • Jeweled movement
  • Easy-to-read tilted dial
  • Axial support
  • 210° swivel arc
  • Automatic reversal
  • Revolution counter
  • Shop hardened case
  • Includes: dial test indicator, shop-hardened case, axial support, contact point case, two dovetail stems, contact point wrench, height gage bar with clamp, three contact points, adapter sleeve.