Learn More
MSC Fowler Test Indicator Kit, 7 Pc, 0.2500 in, 0.0002 in Accuracy
Catalog No. 5000533773
Encompass
This 7-piece Test Indicator Kit provides precise measurements for centering and positioning applications. It features a swivel head indicator style with a maximum measurement of 0.2500 inch and an accuracy of 0.0002 "
  • Designed for centering and positioning tasks.
  • Provides 2D coaxial measurement.
  • Includes a restraining arm for stability.
  • Comes with a coaxial indicator.
  • Supplied with a center point feeler.
  • Contains three curved feelers.
  • Packaged in a protective case.
Catalog No. 50-005-33773 Supplier MSC Supplier No. 35520881
This 7-piece Test Indicator Kit provides precise measurements for centering and positioning applications. It features a swivel head indicator style with a maximum measurement of 0.2500 inch and an accuracy of 0.0002 "
  • Designed for centering and positioning tasks.
  • Provides 2D coaxial measurement.
  • Includes a restraining arm for stability.
  • Comes with a coaxial indicator.
  • Supplied with a center point feeler.
  • Contains three curved feelers.
  • Packaged in a protective case.