Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 3169MSM1P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: FOXA1/1241. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: HepG-2 or MCF-7 cells. Liver, Pancreas, Bladder, Prostate, Colon and Lung and Breast. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant human FOXA1 protein fragment (aa467-614). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 79kDa.Chromosome: 14q21.1. Gene Symbol: FOXA1. Gene ID: 3169. Ensemble: ENSG00000129514. Uniprot: NP_004487.
Catalog No. 50-188-4778