Learn More
Aviva Systems Biology Corp FOXA1 Antibody - N-terminal region (ARP57867_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57867_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FOXA1. Alternate names: HNF3A, MGC33105, TCF3AApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rat
Catalog No. 50-014-62164