Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology FoxA1/HNF3α Monoclonal Antibody, Mouse, IHC578, 100 μL
SDP

Catalog No. 502732733
Encompass

This mouse monoclonal antibody recognizes endogenous levels of total FoxA1/HNF3α protein. The antibody is produced by immunizing animals with a recombinant protein corresponding to human FoxA1/HNF3α. FoxA1 is a transcription factor acting as a pioneer factor that is recruited to distant enhancers to alter chromatin structure for cell type-specific transcription. It is implicated in endoderm-derived organ development and hormone-dependent disorders such as prostate and breast cancers.

  • Host species: Mouse
  • Isotype: IgG1
  • Species reactivity: Human
  • Applications: Immunohistochemistry (paraffin)
  • Recommended dilution for immunohistochemistry (paraffin): 1:800 to 1:3200
  • Supplied in a Tris-based buffer with 1% BSA and less than 0.1% sodium azide
  • Stability: 24 months when stored at 4°C
  • Not for use in diagnostic procedures

Catalog No. 50-273-2733 Supplier Cell Signaling Technology Supplier No. 59197S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This mouse monoclonal antibody recognizes endogenous levels of total FoxA1/HNF3α protein. The antibody is produced by immunizing animals with a recombinant protein corresponding to human FoxA1/HNF3α. FoxA1 is a transcription factor acting as a pioneer factor that is recruited to distant enhancers to alter chromatin structure for cell type-specific transcription. It is implicated in endoderm-derived organ development and hormone-dependent disorders such as prostate and breast cancers.

  • Host species: Mouse
  • Isotype: IgG1
  • Species reactivity: Human
  • Applications: Immunohistochemistry (paraffin)
  • Recommended dilution for immunohistochemistry (paraffin): 1:800 to 1:3200
  • Supplied in a Tris-based buffer with 1% BSA and less than 0.1% sodium azide
  • Stability: 24 months when stored at 4°C
  • Not for use in diagnostic procedures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.