Learn More
Aviva Systems Biology Corp FOXA2 Antibody - N-terminal region (ARP39508_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39508_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against.XA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FOXA2. Alternate names: HNF3B, TCF3BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57064