Learn More
Aviva Systems Biology Corp FOXA2 Antibody - C-terminal region (ARP39509_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39509_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against.XA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FOXA2. Alternate names: HNF3B, TCF3BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57065