Learn More
Aviva Systems Biology Corp Foxa2 Antibody - middle region (ARP37060_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37060_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.xa2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse Foxa2. Alternate names: HNF3-beta, HNF3beta, Hnf-3b, Hnf3b, Tcf-3b, Tcf3bApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-284-576