Learn More
Aviva Systems Biology Corp FOXA3 Antibody - middle region (ARP31945_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31945_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XA3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human FOXA3. Alternate names: FKHH3, HNF3G, TCF3GApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54386