Learn More
OriGene FOXA3 (NM_004497) Human Over-expression Lysate, 20ug
SDP
Catalog No. NC3359310
Encompass_Preferred
FOXA3 HEK293T cell transient overexpression lysate (as WB positive control)
Catalog No. NC3359310 Supplier OriGene Supplier No. LC401428
FOXA3 HEK293T cell transient overexpression lysate (as WB positive control)