Learn More
Aviva Systems Biology Corp FOXB1 Antibody - middle region (ARP35849_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35849_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XB1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FOXB1. Alternate names: FKH5, HFKH-5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-55889