Learn More
Aviva Systems Biology Corp Foxb2 Antibody - C-terminal region (ARP36856_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36856_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.xb2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the c terminal region of mouse Foxb2. Alternate names: Fkh4ApplicationWB. Species reactivity: Cow, Mouse, Rabbit
Catalog No. 50-284-527