Learn More
Aviva Systems Biology Corp FOXC1 Antibody - C-terminal region (ARP32300_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32300_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against.XC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human FOXC1. Alternate names: ARA, IGDA, IHG1, FKHL7, IRID1, RIEG3, FREAC3, FREAC-3ApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-54517