Learn More
Aviva Systems Biology Corp FOXC1 Antibody - C-terminal region (ARP38037_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38037_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XC1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human FOXC1. Alternate names: ARA, FKHL7, FREAC-3, FREAC3, IGDA, IHG1, IRID1, RIEG3ApplicationWB. Species reactivity: Human, Mouse, Rat, Zebrafish
Catalog No. 50-284-709