Learn More
Aviva Systems Biology Corp FOXC1 Antibody - N-terminal region (ARP38036_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38036_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FOXC1. Alternate names: ARA, FKHL7, FREAC3, IGDA, IHG1, IRID1, RIEG3, FREAC-3ApplicationIHC, WB. Species reactivity: Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-56495