Learn More
Aviva Systems Biology Corp FOXD1 Antibody - N-terminal region (ARP36877_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36877_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against.XD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse FOXD1. Alternate names: BF-2, Hfh10, FREAC4, Hfhbf2, AI385632ApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-014-56288