Learn More
Aviva Systems Biology Corp FOXD2 Antibody - N-terminal region (ARP31709_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31709_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against.XD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FOXD2. Alternate names: FKHL17, FREAC9, FREAC-9ApplicationWB. Species reactivity: Cow, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-54316