Learn More
Aviva Systems Biology Corp FOXD3 Antibody - middle region (ARP32561_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32561_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against.XD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FOXD3. Alternate names: AIS1, HFH2, VAMAS2, GenesisApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rat
Catalog No. 50-014-54657